siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107596.1 | NP_001101066.1 | 6 | 1441 | ||
| XM_006231163.1 | XP_006231225.1 | 6 | 1454 | |||
| XM_006231164.2 | XP_006231226.1 | 7 | 1728 | |||
| XM_006231165.2 | XP_006231227.1 | 7 | 2566 | |||
| XM_006231166.2 | XP_006231228.1 | 7 | 1562 | |||
| XM_008760298.1 | XP_008758520.1 | 8 | 1903 | |||
| XM_008760299.1 | XP_008758521.1 | 7 | 1614 | |||

Tmem2
Genomic Map