siRNA Details
| Gene Symbol: Trpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191562.1 | NP_001178491.1 | 4236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpm3| Gene Symbol: Trpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191562.1 | NP_001178491.1 | 4236 | |||