siRNA Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760316.1 | XP_008758538.1 | 7 | 1613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mamdc2| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760316.1 | XP_008758538.1 | 7 | 1613 | ||