siRNA Details
| Gene Symbol: Fam189a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_219909.8 | XP_219909.7 | 10 | 1415 | ||
| GenBank mRNA | BC166504.1 | - | 10 | 1200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189a2| Gene Symbol: Fam189a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_219909.8 | XP_219909.7 | 10 | 1415 | ||
| GenBank mRNA | BC166504.1 | - | 10 | 1200 | ||