siRNA Details
| Gene Symbol: Sec31b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135713.1 | NP_001129185.1 | 13 | 1637 | ||
| XM_006231484.2 | XP_006231546.1 | 13 | 1745 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec31b| Gene Symbol: Sec31b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135713.1 | NP_001129185.1 | 13 | 1637 | ||
| XM_006231484.2 | XP_006231546.1 | 13 | 1745 | |||