siRNA Details
| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113749.1 | NP_001107221.1 | 2 | 310 | ||
| GenBank mRNA | BC158810.1 | - | 2 | 310 | 7368955 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hif1an| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113749.1 | NP_001107221.1 | 2 | 310 | ||
| GenBank mRNA | BC158810.1 | - | 2 | 310 | 7368955 | |