siRNA Details
| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107598.1 | NP_001101068.1 | 2 | 543 | ||
| XM_008760428.1 | XP_008758650.1 | 3 | 464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl43| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107598.1 | NP_001101068.1 | 2 | 543 | ||
| XM_008760428.1 | XP_008758650.1 | 3 | 464 | |||