siRNA Details
| Gene Symbol: Hps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181432.2 | NP_852097.1 | 1 | 348 | ||
| GenBank mRNA | BC086975.1 | - | 1 | 348 | 7193621 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hps6| Gene Symbol: Hps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181432.2 | NP_852097.1 | 1 | 348 | ||
| GenBank mRNA | BC086975.1 | - | 1 | 348 | 7193621 | |