siRNA Details
| Gene Symbol: Tm9sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231413.1 | XP_006231475.1 | 13 | 2001 | ||
| GenBank mRNA | FQ209899.1 | - | 10 | 1116 | ||
| FQ229341.1 | - | 9 | 1025 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm9sf3| Gene Symbol: Tm9sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231413.1 | XP_006231475.1 | 13 | 2001 | ||
| GenBank mRNA | FQ209899.1 | - | 10 | 1116 | ||
| FQ229341.1 | - | 9 | 1025 | |||