siRNA Details
| Gene Symbol: Ch25h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025415.1 | NP_001020586.1 | 1 | 862 | ||
| XM_006231291.2 | XP_006231353.1 | 2 | 3463 | |||
| GenBank mRNA | BC097964.1 | - | 1 | 862 | 7387594 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ch25h| Gene Symbol: Ch25h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025415.1 | NP_001020586.1 | 1 | 862 | ||
| XM_006231291.2 | XP_006231353.1 | 2 | 3463 | |||
| GenBank mRNA | BC097964.1 | - | 1 | 862 | 7387594 | |