siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748666.3 | XP_003748714.1 | 13 | 1649 | ||
| XM_006227693.2 | XP_006227755.1 | 12 | 3305 | |||
| XM_006227695.2 | XP_006227757.1 | 13 | 1363 | |||
| XM_006227696.2 | XP_006227758.1 | 12 | 1282 | |||
| XM_006227697.2 | XP_006227759.1 | 12 | 1568 | |||
| XM_006227698.2 | XP_006227760.1 | 11 | 3230 | |||
| XM_006227699.2 | XP_006227761.1 | 11 | 1128 | |||
| XM_008758644.1 | XP_008756866.1 | 12 | 2484 | |||

L3mbtl3
Genomic Map