siRNA Details
| Gene Symbol: Epb41l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227709.1 | XP_006227771.1 | 4 | 956 | ||
| XM_008758646.1 | XP_008756868.1 | 4 | 1138 | |||
| XM_008758647.1 | XP_008756869.1 | 4 | 1254 | |||
| XM_008758648.1 | XP_008756870.1 | 4 | 859 | |||
| XM_008758649.1 | XP_008756871.1 | 5 | 1094 | |||
| XM_008758650.1 | XP_008756872.1 | 4 | 3941 | |||
| GenBank mRNA | FQ212032.1 | - | 5 | 1052 | ||

Epb41l2
Genomic Map