siRNA Details
| Gene Symbol: Med23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277054.1 | NP_001263983.1 | 23 | 3151 | ||
| XM_008758612.1 | XP_008756834.1 | 24 | 3942 | |||
| XM_008758613.1 | XP_008756835.1 | 22 | 3827 | |||
| XM_008758614.1 | XP_008756836.1 | 24 | 3943 | |||
| GenBank mRNA | BC090022.1 | - | 5 | 813 | ||

Med23
Genomic Map