siRNA Details
| Gene Symbol: Olr1709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000509.1 | NP_001000509.1 | 1 | 486 | ||
| Gene Symbol: Olr1707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001115.1 | NP_001001115.1 | 1 | 486 | ||

Olr1709
Genomic Map