siRNA Details
| Gene Symbol: Olr1733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000510.1 | NP_001000510.1 | 1 | 471 | ||
| Gene Symbol: Olr1739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001423.1 | NP_001001423.1 | 1 | 471 | ||

Olr1733
Genomic Map