siRNA Details
| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212500.3 | NP_997665.1 | 3 | 380 | ||
| GenBank mRNA | BC129081.1 | - | 3 | 285 | 7442774 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnl1| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212500.3 | NP_997665.1 | 3 | 380 | ||
| GenBank mRNA | BC129081.1 | - | 3 | 285 | 7442774 | |