siRNA Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166275.1 | NP_001159747.1 | 11 | 3135 | ||
| GenBank mRNA | BC085955.1 | - | 11 | 3135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mdc1| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166275.1 | NP_001159747.1 | 11 | 3135 | ||
| GenBank mRNA | BC085955.1 | - | 11 | 3135 | ||