siRNA Details
| Gene Symbol: Ly6g6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003691.1 | NP_001003691.1 | 5 | 868 | ||
| XM_006256066.2 | XP_006256128.1 | 5 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6g6f| Gene Symbol: Ly6g6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003691.1 | NP_001003691.1 | 5 | 868 | ||
| XM_006256066.2 | XP_006256128.1 | 5 | 923 | |||