siRNA Details
| Gene Symbol: ZNF282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303481.1 | NP_001290410.1 | 6 | 1173 | ||
| NM_003575.3 | NP_003566.1 | 6 | 1173 | |||
| XM_006716151.3 | XP_006716214.1 | 6 | 1170 | |||
| GenBank mRNA | AK299280.1 | - | 6 | 1085 | ||
| AK316282.1 | - | 6 | 1060 | |||
| AY927504.1 | - | 4 | 440 | |||
| AY927505.1 | - | 5 | 846 | |||
| BC041704.1 | - | 6 | 900 | |||
| BC055287.1 | - | 6 | 963 | |||
| BC073805.1 | - | 6 | 1066 | 6645861 | ||
| D30612.1 | - | 6 | 1186 | |||

ZNF282
Genomic Map