siRNA Details
| Gene Symbol: Anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256193.2 | XP_006256255.1 | 3, 4 | 565 | ||
| XM_006256196.2 | XP_006256258.1 | 3, 4 | 565 | |||
| XM_008772813.1 | XP_008771035.1 | 3, 4 | 565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anks1a| Gene Symbol: Anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256193.2 | XP_006256255.1 | 3, 4 | 565 | ||
| XM_006256196.2 | XP_006256258.1 | 3, 4 | 565 | |||
| XM_008772813.1 | XP_008771035.1 | 3, 4 | 565 | |||