siRNA Details
| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007695.1 | NP_001007696.1 | 9 | 1403 | ||
| XM_006256200.2 | XP_006256262.1 | 9 | 1262 | |||
| GenBank mRNA | BC083899.1 | - | 9 | 1403 | 7133508 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcp11| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007695.1 | NP_001007696.1 | 9 | 1403 | ||
| XM_006256200.2 | XP_006256262.1 | 9 | 1262 | |||
| GenBank mRNA | BC083899.1 | - | 9 | 1403 | 7133508 | |