siRNA Details
| Gene Symbol: Ppil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034188.1 | NP_001029360.1 | 4 | 1250 | ||
| GenBank mRNA | BC099188.1 | - | 4 | 1250 | 7383385 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppil1| Gene Symbol: Ppil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034188.1 | NP_001029360.1 | 4 | 1250 | ||
| GenBank mRNA | BC099188.1 | - | 4 | 1250 | 7383385 | |