siRNA Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012073.1 | NP_001012073.1 | 6 | 504 | ||
| GenBank mRNA | BC087152.1 | - | 6 | 504 | 7132167 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrp1| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012073.1 | NP_001012073.1 | 6 | 504 | ||
| GenBank mRNA | BC087152.1 | - | 6 | 504 | 7132167 | |