siRNA Details
| Gene Symbol: RGD1309594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008351.1 | NP_001008352.1 | 3 | 178 | ||
| XM_006256252.2 | XP_006256314.1 | 3 | 476 | |||
| GenBank mRNA | BC085944.1 | - | 3 | 178 | 7127440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309594| Gene Symbol: RGD1309594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008351.1 | NP_001008352.1 | 3 | 178 | ||
| XM_006256252.2 | XP_006256314.1 | 3 | 476 | |||
| GenBank mRNA | BC085944.1 | - | 3 | 178 | 7127440 | |