siRNA Details
| Gene Symbol: Bcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228091.9 | XP_228091.6 | 14 | 2971 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcr| Gene Symbol: Bcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_228091.9 | XP_228091.6 | 14 | 2971 | ||