siRNA Details
| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107634.1 | NP_001101104.1 | 4 | 669 | ||
| XM_008772985.1 | XP_008771207.1 | 3 | 284 | |||
| XM_008772986.1 | XP_008771208.1 | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk19| Gene Symbol: Cdk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107634.1 | NP_001101104.1 | 4 | 669 | ||
| XM_008772985.1 | XP_008771207.1 | 3 | 284 | |||
| XM_008772986.1 | XP_008771208.1 | 1 | 23 | |||