siRNA Details
| Gene Symbol: Rev3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083966.1 | NP_001077435.1 | 13 | 4347 | ||
| XM_006256524.1 | XP_006256586.1 | 13 | 4352 | |||
| XM_006256525.2 | XP_006256587.1 | 14 | 4110 | |||
| XM_006256527.2 | XP_006256589.1 | 13 | 4352 | |||
| XM_008772987.1 | XP_008771209.1 | 13 | 4413 | |||
| XM_008772988.1 | XP_008771210.1 | 13 | 4352 | |||

Rev3l
Genomic Map