siRNA Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107636.1 | NP_001101106.1 | 12 | 1891 | ||
| XM_006256449.2 | XP_006256511.1 | 13 | 2157 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts14| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107636.1 | NP_001101106.1 | 12 | 1891 | ||
| XM_006256449.2 | XP_006256511.1 | 13 | 2157 | |||