siRNA Details
| Gene Symbol: Fig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047096.1 | NP_001040561.1 | 18 | 2046 | ||
| XM_006256540.2 | XP_006256602.1 | 17 | 2885 | |||
| XM_006256541.2 | XP_006256603.1 | 16 | 1821 | |||
| GenBank mRNA | DQ666853.1 | - | 18 | 2046 | ||
| FQ213468.1 | - | 1 | 65 | |||

Fig4
Genomic Map