siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107638.1 | NP_001101108.1 | 8 | 1255 | ||
| XM_006256572.2 | XP_006256634.1 | 8 | 1552 | |||
| XM_008772989.1 | XP_008771211.1 | 4 | 1436 | |||
| XM_008772990.1 | XP_008771212.1 | 4 | 852 | |||

Scml4
Genomic Map