siRNA Details
| Gene Symbol: Rtn4ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107644.3 | NP_001101114.1 | 2 | 711 | ||
| XM_006256589.2 | XP_006256651.1 | 2 | 732 | |||
| XM_006256590.2 | XP_006256652.1 | 2 | 732 | |||
| XM_008772999.1 | XP_008771221.1 | 3 | 400 | |||
| GenBank mRNA | BC082054.1 | - | 2 | 732 | ||
| BC166465.1 | - | 2 | 715 | 9030981 | ||

Rtn4ip1
Genomic Map