siRNA Details
| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007797.1 | NP_001007798.1 | 5 | 858 | ||
| GenBank mRNA | AY383703.1 | - | 5 | 858 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arrdc3| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007797.1 | NP_001007798.1 | 5 | 858 | ||
| GenBank mRNA | AY383703.1 | - | 5 | 858 | ||