siRNA Details
| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013156.1 | NP_001013174.1 | 1 | 177 | ||
| XM_006231841.2 | XP_006231903.1 | 2 | 279 | |||
| GenBank mRNA | BC083695.1 | - | 1 | 177 | 7189545 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc9| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013156.1 | NP_001013174.1 | 1 | 177 | ||
| XM_006231841.2 | XP_006231903.1 | 2 | 279 | |||
| GenBank mRNA | BC083695.1 | - | 1 | 177 | 7189545 | |