siRNA Details
| Gene Symbol: Ptcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107648.1 | NP_001101118.1 | 9 | 858 | ||
| XM_006231847.2 | XP_006231909.1 | 10 | 929 | |||
| XM_008760675.1 | XP_008758897.1 | 9 | 2119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptcd2| Gene Symbol: Ptcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107648.1 | NP_001101118.1 | 9 | 858 | ||
| XM_006231847.2 | XP_006231909.1 | 10 | 929 | |||
| XM_008760675.1 | XP_008758897.1 | 9 | 2119 | |||