siRNA Details
| Gene Symbol: Mast4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760713.1 | XP_008758935.1 | 1638 | |||
| Gene Symbol: NEWGENE_1310139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760704.1 | XP_008758926.1 | 14 | 2986 | ||
| XM_008760705.1 | XP_008758927.1 | 14 | 2986 | |||

Mast4
Genomic Map