siRNA Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231909.2 | XP_006231971.1 | 5 | 973 | ||
| XM_008760730.1 | XP_008758952.1 | 2 | 3433 | |||
| GenBank mRNA | AY325185.1 | - | 3 | 389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim6| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231909.2 | XP_006231971.1 | 5 | 973 | ||
| XM_008760730.1 | XP_008758952.1 | 2 | 3433 | |||
| GenBank mRNA | AY325185.1 | - | 3 | 389 | ||