siRNA Details
| Gene Symbol: Ercc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107650.2 | NP_001101120.2 | 9 | 814 | ||
| XM_008760726.1 | XP_008758948.1 | 9 | 876 | |||
| XM_008760727.1 | XP_008758949.1 | 9 | 879 | |||
| XM_008760728.1 | XP_008758950.1 | 8 | 724 | |||
| GenBank mRNA | BC089220.1 | - | 9 | 729 | ||
| BC161924.1 | - | 8 | 689 | 8370925 | ||

Ercc8
Genomic Map