siRNA Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035251.1 | NP_001030328.1 | 3 | 358 | ||
| XM_006231929.2 | XP_006231991.1 | 4 | 485 | |||
| XM_006231930.2 | XP_006231992.1 | 5 | 549 | |||
| XM_006231931.2 | XP_006231993.1 | 4 | 526 | |||
| XM_006231932.2 | XP_006231994.1 | 4 | 616 | |||
| XM_006231933.2 | XP_006231995.1 | 4 | 575 | |||
| XM_006231934.2 | XP_006231996.1 | 3 | 2560 | |||
| XM_006231936.2 | XP_006231998.1 | 3 | 2545 | |||
| XM_006231937.2 | XP_006231999.1 | 3 | 326 | |||
| XM_006231939.2 | XP_006232001.1 | 3 | 238 | |||
| GenBank mRNA | BC092662.1 | - | 2 | 220 | ||
| BC105869.1 | - | 3 | 358 | 7442690 | ||
| JQ013729.1 | - | 3 | 409 | |||

Slc38a9
Genomic Map