siRNA Details
| Gene Symbol: Slc45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107653.1 | NP_001101123.1 | 3 | 936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc45a2| Gene Symbol: Slc45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107653.1 | NP_001101123.1 | 3 | 936 | ||