siRNA Details
| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012077.1 | NP_001012077.1 | 15 | 1742 | ||
| GenBank mRNA | BC089876.1 | - | 15 | 1742 | 7325487 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr12| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012077.1 | NP_001012077.1 | 15 | 1742 | ||
| GenBank mRNA | BC089876.1 | - | 15 | 1742 | 7325487 | |