siRNA Details
| Gene Symbol: Ankrd33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191565.1 | NP_001178494.1 | 4 | 1191 | ||
| GenBank mRNA | FQ232932.1 | - | 4 | 1380 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd33b| Gene Symbol: Ankrd33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191565.1 | NP_001178494.1 | 4 | 1191 | ||
| GenBank mRNA | FQ232932.1 | - | 4 | 1380 | ||