siRNA Details
| Gene Symbol: Cmbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008770.3 | NP_001008770.1 | 3 | 329 | ||
| XM_006232097.2 | XP_006232159.1 | 3 | 2440 | |||
| GenBank mRNA | AY325197.1 | - | 3 | 331 | ||
| BC088459.1 | - | 2 | 313 | 7127481 | ||
| BF556107.1 | - | 3 | 336 | |||
| FQ209709.1 | - | 3 | 487 | |||
| FQ213936.1 | - | 3 | 339 | |||
| FQ215832.1 | - | 2 | 331 | |||
| FQ219291.1 | - | 3 | 418 | |||
| FQ219760.1 | - | 3 | 342 | |||

Cmbl
Genomic Map