siRNA Details
| Gene Symbol: Slc10a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025280.1 | NP_001020451.1 | 1 | 752 | ||
| GenBank mRNA | DQ074435.1 | - | 1 | 752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc10a5| Gene Symbol: Slc10a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025280.1 | NP_001020451.1 | 1 | 752 | ||
| GenBank mRNA | DQ074435.1 | - | 1 | 752 | ||