siRNA Details
| Gene Symbol: Ttc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107666.2 | NP_001101136.2 | 12 | 1665 | ||
| XM_006232257.2 | XP_006232319.1 | 11 | 1559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc14| Gene Symbol: Ttc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107666.2 | NP_001101136.2 | 12 | 1665 | ||
| XM_006232257.2 | XP_006232319.1 | 11 | 1559 | |||