siRNA Details
| Gene Symbol: Ccdc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107667.1 | NP_001101137.1 | 10 | 1216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc39| Gene Symbol: Ccdc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107667.1 | NP_001101137.1 | 10 | 1216 | ||