siRNA Details
| Gene Symbol: Qrfpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198199.1 | NP_937842.1 | 1 | 193 | ||
| GenBank mRNA | AB109630.1 | - | 1 | 193 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qrfpr| Gene Symbol: Qrfpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198199.1 | NP_937842.1 | 1 | 193 | ||
| GenBank mRNA | AB109630.1 | - | 1 | 193 | ||
Set Membership: |
|