siRNA Details
| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191705.1 | NP_001178634.1 | 4479 | |||
| XM_006232292.2 | XP_006232354.1 | 5564 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat4| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191705.1 | NP_001178634.1 | 4479 | |||
| XM_006232292.2 | XP_006232354.1 | 5564 | ||||