siRNA Details
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191705.1 | NP_001178634.1 | 4729 | |||
XM_006232292.2 | XP_006232354.1 | 5814 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191705.1 | NP_001178634.1 | 4729 | |||
XM_006232292.2 | XP_006232354.1 | 5814 |