siRNA Details
| Gene Symbol: RGD1305938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134548.1 | NP_001128020.1 | 2 | 201 | ||
| XM_008760770.1 | XP_008758992.1 | 2 | 446 | |||
| GenBank mRNA | FQ211901.1 | - | 2 | 194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305938| Gene Symbol: RGD1305938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134548.1 | NP_001128020.1 | 2 | 201 | ||
| XM_008760770.1 | XP_008758992.1 | 2 | 446 | |||
| GenBank mRNA | FQ211901.1 | - | 2 | 194 | ||