siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100698.1 | NP_001094168.1 | 4 | 647 | ||
| XM_006231960.2 | XP_006232022.1 | 4 | 642 | |||
| XM_006231961.2 | XP_006232023.1 | 5 | 871 | |||
| XM_006231962.2 | XP_006232024.1 | 5 | 697 | |||
| XM_006231963.2 | XP_006232025.1 | 5 | 720 | |||
| XM_006231965.2 | XP_006232027.1 | 5 | 885 | |||
| XM_008760772.1 | XP_008758994.1 | 4 | 1074 | |||
| GenBank mRNA | BC161974.1 | - | 4 | 647 | 9029363 | |

Zfp131
Genomic Map